Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmod2
Entrez Gene ID
50876
Gene Name
tropomodulin 2
Gene Aliases
N-Tmod, NTMOD
Chromosome Location
Chr.9: 75565621 - 75611348 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016711.3 NP_057920.2 1 - 2 306 56
GenBank mRNA AF177170.1 - 1 - 2 255 56
AK018223.1 - 1 - 2 103 56
AK079077.1 - 1 - 2 150 56
AK164178.1 - 1 - 2 152 56

View Full Product Details