The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205253.1 | NP_001192182.1 | 3 - 4 | 1244 | 103 |
NM_019576.2 | NP_062522.1 | 4 - 5 | 1387 | 103 | |
GenBank mRNA | AB039946.1 | - | 3 - 4 | 1215 | 103 |
AK049636.1 | - | 4 - 5 | 1350 | 103 | |
AK052555.1 | - | 3 - 4 | 1185 | 103 | |
BC119603.2 | - | 3 - 4 | 1215 | 103 |