The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019956.1 | NP_064340.1 | 8 - 9 | 1411 | 61 |
GenBank mRNA | AB033744.1 | - | 8 - 9 | 1411 | 61 |
AB100413.1 | - | 8 - 9 | 1411 | 61 | |
AK003704.1 | - | 8 - 9 | 1423 | 61 | |
AK004268.1 | - | 2 - 3 | 220 | 61 | |
AY033497.1 | - | 8 - 9 | 1411 | 61 | |
BC125346.1 | - | 8 - 9 | 1406 | 61 | |
BC125348.1 | - | 8 - 9 | 1406 | 61 |