The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199118.1 | NP_001186047.1 | 11 - 12 | 1772 | 100 |
NM_022883.3 | NP_075021.1 | 11 - 12 | 1849 | 100 | |
GenBank mRNA | AF286724.1 | - | 11 - 12 | 1786 | 100 |
AK049363.1 | - | 11 - 12 | 1765 | 100 | |
AK163833.1 | - | 11 - 12 | 1764 | 100 | |
AK220228.1 | - | 11 - 12 | 1849 | 100 | |
BC026729.1 | - | 11 - 12 | 1756 | 100 | |
BC117882.1 | - | 10 - 11 | 1713 | 100 | |
BC117883.1 | - | 10 - 11 | 1713 | 100 |