The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177955.1 | NP_001171426.1 | 5 - 6 | 1163 | 69 |
NM_001177956.1 | NP_001171427.1 | 5 - 6 | 1163 | 69 | |
NM_001177957.1 | NP_001171428.1 | 5 - 6 | 1163 | 69 | |
NM_001177958.1 | NP_001171429.1 | 4 - 5 | 1043 | 69 | |
NM_001177959.1 | NP_001171430.1 | 4 - 5 | 1043 | 69 | |
NM_001177960.1 | NP_001171431.1 | 4 - 5 | 1043 | 69 | |
NM_001177961.1 | NP_001171432.1 | 4 - 5 | 708 | 69 | |
NM_001177962.1 | NP_001171433.1 | 4 - 5 | 708 | 69 | |
NM_023122.3 | NP_075611.1 | 4 - 5 | 708 | 69 | |
GenBank mRNA | AF254869.1 | - | 4 - 5 | 904 | 69 |
AF254870.1 | - | 5 - 6 | 1024 | 69 | |
AF254871.1 | - | 4 - 5 | 679 | 69 | |
AF254872.1 | - | 4 - 5 | 904 | 69 | |
AF254873.1 | - | 4 - 5 | 904 | 69 | |
AF254874.1 | - | 5 - 6 | 1024 | 69 | |
AF254875.1 | - | 5 - 6 | 1024 | 69 | |
AF254876.1 | - | 4 - 5 | 679 | 69 | |
AF254877.1 | - | 4 - 5 | 679 | 69 | |
AF254878.1 | - | 5 - 6 | 1051 | 69 | |
AF254879.1 | - | 6 - 7 | 1171 | 69 | |
AK016567.1 | - | 4 - 5 | 633 | 69 | |
AK132778.1 | - | 4 - 5 | 695 | 69 | |
AK140714.1 | - | 4 - 5 | 723 | 69 | |
BC003912.1 | - | 3 - 4 | 491 | 69 | |
BC025165.1 | - | 4 - 5 | 669 | 69 | |
S65737.1 | - | 4 - 5 | 700 | 69 |