Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dazap1
Entrez Gene ID
70248
Gene Name
DAZ associated protein 1
Gene Aliases
2410042M16Rik, mPrrp
Chromosome Location
Chr.10: 80261471 - 80288413 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122604.1 NP_001116076.1 6 - 7 626 96
NM_001122605.1 NP_001116077.1 6 - 7 623 96
NM_133188.2 NP_573451.2 6 - 7 623 96
GenBank mRNA AF225910.1 - 6 - 7 517 96
AK148023.1 - 6 - 7 501 96
AK152314.1 - 6 - 7 593 96
AK152345.1 - 6 - 7 593 96
BC049355.1 - 6 - 7 543 96
BC058775.1 - 6 - 7 537 96

View Full Product Details