The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290398.1 | NP_001277327.1 | 13 - 14 | 1857 | 155 |
NM_133196.6 | NP_573459.1 | 13 - 14 | 1935 | 155 | |
GenBank mRNA | AF317552.1 | - | 13 - 14 | 1903 | 155 |
AK032817.1 | - | 13 - 14 | 1917 | 155 | |
AK047255.1 | - | 1 - 2 | 841 | 155 | |
AK049971.1 | - | 13 - 14 | 1848 | 155 | |
AK088260.1 | - | 13 - 14 | 1763 | 155 | |
AK147438.1 | - | 11 - 12 | 4929 | 155 | |
EU616680.1 | - | 15 - 16 | 2053 | 155 | |
EU616681.1 | - | 13 - 14 | 1825 | 155 | |
EU616682.1 | - | 15 - 16 | 1975 | 155 |