The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007998.6 | NP_032024.2 | 10 - 11 | 1233 | 63 |
GenBank mRNA | AK004718.2 | - | 10 - 11 | 1234 | 63 |
AK018738.2 | - | 10 - 11 | 1236 | 63 | |
AK084843.1 | - | 10 - 11 | 1131 | 63 | |
AK150685.1 | - | 10 - 11 | 1235 | 63 | |
AK159652.1 | - | 10 - 11 | 1266 | 63 | |
BC094548.1 | - | 10 - 11 | 1206 | 63 | |
M59288.1 | - | 9 - 10 | 1070 | 63 |