The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045489.1 | NP_001038954.1 | 1 - 2 | 165 | 53 |
NM_008594.2 | NP_032620.2 | 1 - 2 | 165 | 53 | |
GenBank mRNA | AB021130.2 | - | 1 - 2 | 72 | 53 |
AB025280.1 | - | 1 - 2 | 72 | 53 | |
AK087555.1 | - | 1 - 2 | 154 | 53 | |
AK089211.1 | - | 1 - 2 | 131 | 53 | |
AK145183.1 | - | 1 - 2 | 129 | 53 | |
AK148371.1 | - | 1 - 2 | 154 | 53 | |
AK152088.1 | - | 1 - 2 | 154 | 53 | |
AK155047.1 | - | 1 - 2 | 163 | 53 | |
AK169997.1 | - | 1 - 2 | 129 | 53 | |
AK170935.1 | - | 1 - 2 | 129 | 53 | |
AK171143.1 | - | 1 - 2 | 129 | 53 | |
AK209209.1 | - | 1 - 2 | 78 | 53 | |
BC003892.1 | - | 1 - 2 | 109 | 53 | |
BC003904.1 | - | 1 - 2 | 104 | 53 | |
BC018577.1 | - | 1 - 2 | 102 | 53 | |
M38337.1 | - | 1 - 2 | 117 | 53 | |
Y11684.2 | - | 1 - 2 | 72 | 53 |