The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045489.1 | NP_001038954.1 | 7 - 8 | 1082 | 70 |
NM_008594.2 | NP_032620.2 | 8 - 9 | 1193 | 70 | |
GenBank mRNA | AB021130.2 | - | 8 - 9 | 1100 | 70 |
AB025280.1 | - | 7 - 8 | 989 | 70 | |
AK087555.1 | - | 7 - 8 | 1072 | 70 | |
AK089211.1 | - | 7 - 8 | 1048 | 70 | |
AK148371.1 | - | 7 - 8 | 1071 | 70 | |
AK152088.1 | - | 8 - 9 | 1182 | 70 | |
AK155047.1 | - | 8 - 9 | 1191 | 70 | |
AK169997.1 | - | 7 - 8 | 1046 | 70 | |
AK170935.1 | - | 7 - 8 | 1046 | 70 | |
AK171143.1 | - | 8 - 9 | 1157 | 70 | |
AK190819.1 | - | 2 - 3 | 212 | 70 | |
AK194123.1 | - | 2 - 3 | 212 | 70 | |
AK194145.1 | - | 2 - 3 | 212 | 70 | |
BC003892.1 | - | 7 - 8 | 1026 | 70 | |
BC003904.1 | - | 7 - 8 | 1021 | 70 | |
BC018577.1 | - | 7 - 8 | 1019 | 70 | |
BC037475.1 | - | 4 - 5 | 1408 | 70 | |
M38337.1 | - | 8 - 9 | 1145 | 70 | |
Y11684.2 | - | 7 - 8 | 989 | 70 |