The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008802.3 | NP_032828.2 | 1 - 2 | 193 | 105 |
GenBank mRNA | AK036517.1 | - | 1 - 2 | 151 | 105 |
AK168957.1 | - | 1 - 2 | 155 | 105 | |
CV558025.1 | - | 1 - 2 | 193 | 105 | |
U68171.1 | - | 1 - 2 | 141 | 105 |