The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008940.2 | NP_032966.1 | 3 - 4 | 974 | 78 |
GenBank mRNA | AB074296.1 | - | 1 - 2 | 236 | 78 |
BC055895.1 | - | 3 - 4 | 488 | 78 | |
D30785.1 | - | 3 - 4 | 974 | 78 |