The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037726.1 | NP_001032815.1 | 4 - 5 | 613 | 103 |
NM_009952.2 | NP_034082.1 | 6 - 7 | 777 | 103 | |
NM_133828.2 | NP_598589.2 | 5 - 6 | 735 | 103 | |
GenBank mRNA | AF448507.1 | - | 6 - 7 | 653 | 103 |
AF448508.1 | - | - | 1305 | 103 | |
AK042595.1 | - | 5 - 6 | 708 | 103 | |
AK044992.1 | - | 5 - 6 | 550 | 103 | |
AK044993.1 | - | 5 - 6 | 550 | 103 | |
AK079223.1 | - | 5 - 6 | 731 | 103 | |
BC021649.1 | - | 5 - 6 | 668 | 103 | |
M95106.1 | - | 6 - 7 | 687 | 103 | |
X92497.1 | - | 4 - 5 | 590 | 103 |