The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016684.2 | NP_057893.2 | 2 - 3 | 523 | 73 |
GenBank mRNA | AK075886.1 | - | 2 - 3 | 517 | 73 |
AK129138.1 | - | 2 - 3 | 523 | 73 | |
AK139837.1 | - | 3 - 4 | 846 | 73 | |
AK142005.1 | - | 2 - 3 | 502 | 73 | |
BC027041.1 | - | 1 - 2 | 481 | 73 | |
U62908.1 | - | 1 - 2 | 832 | 73 |