Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hps1
Entrez Gene ID
192236
Gene Name
Hermansky-Pudlak syndrome 1 homolog (human)
Gene Aliases
6030422N11Rik, BB405864, Gm21361, Hps, ep
Chromosome Location
Chr.19: 42755191 - 42780437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019424.2 NP_062297.1 9 - 10 1078 59
GenBank mRNA AK031397.1 - 9 - 10 1092 59
AK053974.1 - 7 - 8 910 59
AK154994.1 - 9 - 10 1267 59
AK171225.1 - 9 - 10 1054 59
AK181298.1 - 2 - 3 185 59
AK183600.1 - 2 - 3 185 59
BC046329.1 - 7 - 8 899 59
BC058332.1 - 9 - 10 1061 59
BC089331.1 - 9 - 10 1061 59
U78315.1 - 8 - 9 1013 59
U97149.1 - 8 - 9 1027 59

View Full Product Details