Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hps1
Entrez Gene ID
192236
Gene Name
Hermansky-Pudlak syndrome 1 homolog (human)
Gene Aliases
6030422N11Rik, BB405864, Gm21361, Hps, ep
Chromosome Location
Chr.19: 42755191 - 42780437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019424.2 NP_062297.1 18 - 19 2064 62
GenBank mRNA AK053974.1 - 16 - 17 1897 62
AK154994.1 - 18 - 19 2253 62
AK171225.1 - 18 - 19 2040 62
AK207948.1 - 1 - 2 30 62
U78315.1 - 17 - 18 1999 62
U97149.1 - 17 - 18 2013 62

View Full Product Details