Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anks3
Entrez Gene ID
72615
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
Gene Aliases
2700067D09Rik, C81345, mKIAA1977
Chromosome Location
Chr.16: 4941415 - 4964327 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028301.4 NP_082577.2 7 - 8 990 93
GenBank mRNA AK012494.1 - 7 - 8 939 93
AK089037.1 - 7 - 8 965 93
AK129471.1 - 7 - 8 934 93
BC050929.1 - 7 - 8 926 93
BC051143.1 - 7 - 8 957 93

View Full Product Details