Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsga13
Entrez Gene ID
116732
Gene Name
testis specific gene A13
Gene Aliases
1700023D02Rik, Vme1
Chromosome Location
Chr.6: 30896980 - 30925154 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054073.2 NP_473414.1 5 - 6 612 90
GenBank mRNA AF294348.1 - 5 - 6 580 90
AK006265.1 - 5 - 6 613 90
BC125597.1 - 4 - 5 530 90
BC125599.1 - 4 - 5 530 90

View Full Product Details