The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012450.1 | NP_001012453.1 | 6 - 7 | 930 | 61 |
NM_001012451.1 | NP_001012454.1 | 5 - 6 | 641 | 61 | |
NM_080471.3 | NP_536719.2 | 5 - 6 | 770 | 61 | |
GenBank mRNA | AK133551.1 | - | 4 - 5 | 657 | 61 |
AK173070.1 | - | 5 - 6 | 681 | 61 | |
AY026320.2 | - | 4 - 5 | 562 | 61 | |
BC065177.1 | - | 6 - 7 | 880 | 61 |