Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd6
Entrez Gene ID
140577
Gene Name
ankyrin repeat domain 6
Gene Aliases
-
Chromosome Location
Chr.4: 32804027 - 32950884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012450.1 NP_001012453.1 14 - 15 1883 61
NM_001012451.1 NP_001012454.1 13 - 14 1594 61
NM_080471.3 NP_536719.2 13 - 14 1723 61
GenBank mRNA AK133551.1 - 12 - 13 1610 61
AK173070.1 - 13 - 14 1634 61
AY026320.2 - 12 - 13 1515 61
BC065177.1 - 14 - 15 1833 61

View Full Product Details