Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tram2
Entrez Gene ID
170829
Gene Name
translocating chain-associating membrane protein 2
Gene Aliases
C330003D03Rik
Chromosome Location
Chr.1: 21001397 - 21079225 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177409.3 NP_803128.1 2 - 3 292 96
GenBank mRNA AK082751.1 - 2 - 3 293 96
AY029530.1 - 2 - 3 229 96
BC018212.1 - 2 - 3 277 96

View Full Product Details