The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133765.4 | NP_598526.3 | 8 - 9 | 1348 | 79 |
GenBank mRNA | AB041586.1 | - | 8 - 9 | 1197 | 79 |
AK009859.1 | - | 5 - 6 | 790 | 79 | |
AK140412.1 | - | 8 - 9 | 1349 | 79 | |
AK163641.1 | - | 8 - 9 | 1317 | 79 | |
BC026929.1 | - | 8 - 9 | 1333 | 79 |