Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Unc45a
Entrez Gene ID
101869
Gene Name
unc-45 homolog A (C. elegans)
Gene Aliases
AW538196
Chromosome Location
Chr.7: 80325292 - 80341000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133952.2 NP_598713.2 5 - 6 619 60
GenBank mRNA AK028912.1 - 5 - 6 605 60
AK047292.1 - 5 - 6 566 60
AK047476.1 - 5 - 6 566 60
AK088034.1 - 5 - 6 571 60
AK166809.1 - 5 - 6 602 60
BC004717.1 - 5 - 6 608 60

View Full Product Details