Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smek2
Entrez Gene ID
104570
Gene Name
SMEK homolog 2, suppressor of mek1 (Dictyostelium)
Gene Aliases
AW011752, AW557776, mKIAA1387
Chromosome Location
Chr.11: 29167939 - 29220797 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134034.2 NP_598795.2 8 - 9 1755 128
GenBank mRNA AK037253.1 - 8 - 9 1687 128
AK041513.1 - 8 - 9 1700 128
AK044742.1 - 8 - 9 1698 128
AK090014.1 - 8 - 9 1731 128
AK129344.1 - 8 - 9 1666 128
AK132660.1 - 8 - 9 1277 128
AK165940.1 - 8 - 9 1680 128
AK166627.1 - 8 - 9 1682 128
BC006870.1 - 8 - 9 1666 128

View Full Product Details