Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Satb2
Entrez Gene ID
212712
Gene Name
special AT-rich sequence binding protein 2
Gene Aliases
mKIAA1034
Chromosome Location
Chr.1: 56793981 - 56978681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139146.2 NP_631885.1 9 - 10 1827 101
GenBank mRNA AF319623.1 - 9 - 10 1827 101
AJ606045.1 - 8 - 9 1545 101
AK129270.1 - 8 - 9 2818 101
BC138625.1 - 8 - 9 1578 101
BC138626.1 - 8 - 9 1578 101

View Full Product Details