Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Satb2
Entrez Gene ID
212712
Gene Name
special AT-rich sequence binding protein 2
Gene Aliases
mKIAA1034
Chromosome Location
Chr.1: 56793981 - 56978681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139146.2 NP_631885.1 10 - 11 2023 74
GenBank mRNA AF319623.1 - 10 - 11 2023 74
AJ606045.1 - 9 - 10 1741 74
AK129270.1 - 9 - 10 3014 74
BC138625.1 - 9 - 10 1774 74
BC138626.1 - 9 - 10 1774 74

View Full Product Details