Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp45
Entrez Gene ID
77593
Gene Name
ubiquitin specific petidase 45
Gene Aliases
3110003C05Rik, 4930550B20Rik, AI843191, Gcap7
Chromosome Location
Chr.4: 21767161 - 21837872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290425.1 NP_001277354.1 15 - 16 2076 63
NM_152825.2 NP_690038.1 16 - 17 2220 63
GenBank mRNA AK019756.1 - 17 - 18 2336 63
AK036903.1 - 3 - 4 636 63
AK088073.1 - 15 - 16 2059 63
BC027768.1 - 16 - 17 2203 63
BM227454.2 - 2 - 3 63

View Full Product Details