The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024440.2 | NP_077760.1 | 4 - 5 | 349 | 68 |
GenBank mRNA | AB047555.1 | - | 4 - 5 | 349 | 68 |
AK007948.1 | - | 4 - 5 | 335 | 68 | |
AK172429.1 | - | 4 - 5 | 335 | 68 | |
BC004729.1 | - | 4 - 5 | 332 | 68 | |
BC099615.1 | - | 4 - 5 | 329 | 68 | |
BC115466.1 | - | 4 - 5 | 344 | 68 | |
BC115467.1 | - | 4 - 5 | 344 | 68 |