Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl28
Entrez Gene ID
66689
Gene Name
kelch-like 28
Gene Aliases
2810440N09Rik, 4122402F11Rik, 4931401E10Rik, Btbd5
Chromosome Location
Chr.12: 64942198 - 64965605 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025707.3 NP_079983.1 1 - 2 129 74
GenBank mRNA AK002637.1 - 1 - 2 96 74
AK012778.2 - 1 - 2 109 74
AK013278.1 - 1 - 2 107 74
AK014466.1 - 1 - 2 130 74
BC018469.1 - 1 - 2 87 74
BC037017.1 - 1 - 2 102 74

View Full Product Details