Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx31
Entrez Gene ID
66696
Gene Name
sorting nexin 31
Gene Aliases
4631426E05Rik
Chromosome Location
Chr.15: 36504062 - 36555614 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025712.4 NP_079988.3 1 - 2 126 102
GenBank mRNA AK014536.1 - 1 - 2 128 102
AK034196.1 - 1 - 2 128 102
AK035226.1 - 1 - 2 127 102
BC061007.1 - 1 - 2 122 102

View Full Product Details