Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snx31
Entrez Gene ID
66696
Gene Name
sorting nexin 31
Gene Aliases
4631426E05Rik
Chromosome Location
Chr.15: 36504062 - 36555614 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025712.4 NP_079988.3 2 - 3 206 79
GenBank mRNA AK014536.1 - 2 - 3 208 79
AK034196.1 - 2 - 3 208 79
AK035226.1 - 2 - 3 207 79
BC061007.1 - 2 - 3 202 79

View Full Product Details