The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077348.1 | NP_001070816.1 | 3 - 4 | 470 | 108 |
NM_025874.3 | NP_080150.2 | 4 - 5 | 416 | 108 | |
GenBank mRNA | AK010202.1 | - | 4 - 5 | 416 | 108 |
AK075872.1 | - | 4 - 5 | 413 | 108 | |
AY919875.1 | - | 3 - 4 | 429 | 108 | |
BC024138.1 | - | 4 - 5 | 396 | 108 | |
DQ473305.1 | - | 4 - 5 | 396 | 108 |