Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zcchc18
Entrez Gene ID
66995
Gene Name
zinc finger, CCHC domain containing 18
Gene Aliases
1500031H04Rik, Sizn2
Chromosome Location
Chr.X: 136992781 - 136996923 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025893.2 NP_080169.2 1 - 2 200 74
GenBank mRNA AK005317.1 - 1 - 2 178 74

View Full Product Details