The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026012.2 | NP_080288.1 | 4 - 5 | 562 | 61 |
GenBank mRNA | AF534394.1 | - | 3 - 4 | 374 | 61 |
AK007483.1 | - | 2 - 3 | 219 | 61 | |
AK160222.1 | - | 4 - 5 | 61 | ||
BC069856.1 | - | 4 - 5 | 562 | 61 |