The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026192.3 | NP_080468.1 | 14 - 15 | 2055 | 61 |
GenBank mRNA | AK007393.1 | - | 14 - 15 | 1946 | 61 |
AK018734.2 | - | 14 - 15 | 1943 | 61 | |
AK035598.1 | - | 14 - 15 | 1967 | 61 | |
AK220400.1 | - | 14 - 15 | 1968 | 61 | |
AY131329.1 | - | 13 - 14 | 1921 | 61 | |
BC054783.1 | - | 14 - 15 | 1949 | 61 |