The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163035.1 | NP_001156507.1 | 5 - 6 | 869 | 148 |
NM_026421.3 | NP_080697.2 | 5 - 6 | 869 | 148 | |
NR_027990.1 | 5 - 6 | 840 | 148 | ||
GenBank mRNA | AK009954.1 | - | 5 - 6 | 852 | 148 |
AK079062.1 | - | 5 - 6 | 863 | 148 | |
AK080714.1 | - | 5 - 6 | 820 | 148 | |
AK082747.1 | - | 5 - 6 | 844 | 148 | |
AK152021.1 | - | 5 - 6 | 844 | 147 | |
AK167575.1 | - | 5 - 6 | 880 | 148 | |
AK168696.1 | - | 5 - 6 | 738 | 148 | |
BC021429.1 | - | 5 - 6 | 796 | 148 |