Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem86a
Entrez Gene ID
67893
Gene Name
transmembrane protein 86A
Gene Aliases
1810054O13Rik, AI414959, AW742698
Chromosome Location
Chr.7: 47050640 - 47054776 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026436.3 NP_080712.1 1 - 2 159 57
GenBank mRNA AK007864.1 - 1 - 2 160 57
AK075589.1 - 1 - 2 133 57
BC056619.1 - 1 - 2 126 57

View Full Product Details