The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271433.1 | NP_001258362.1 | 8 - 9 | 928 | 68 |
NM_001271434.1 | NP_001258363.1 | 8 - 9 | 930 | 68 | |
NM_001271436.1 | NP_001258365.1 | 7 - 8 | 742 | 68 | |
NM_001271437.1 | NP_001258366.1 | 7 - 8 | 737 | 68 | |
NM_026897.3 | NP_081173.1 | 7 - 8 | 913 | 68 | |
GenBank mRNA | AK005274.1 | - | 7 - 8 | 909 | 68 |
AK012748.1 | - | 8 - 9 | 927 | 68 | |
AK021220.1 | - | 7 - 8 | 744 | 68 | |
AK034798.1 | - | 8 - 9 | 905 | 68 | |
AK154248.1 | - | 7 - 8 | 718 | 68 | |
AK185790.1 | - | 2 - 3 | 108 | 68 | |
BC005586.1 | - | 7 - 8 | 1076 | 68 | |
BC030466.1 | - | 8 - 9 | 913 | 68 | |
BC083322.1 | - | 8 - 9 | 867 | 68 |