Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan2
Entrez Gene ID
70747
Gene Name
tetraspanin 2
Gene Aliases
6330415F13Rik, B230119D02Rik, tspan-2
Chromosome Location
Chr.3: 102734770 - 102772310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243132.1 NP_001230061.1 7 - 8 630 147
NM_027533.3 NP_081809.2 7 - 8 825 147
GenBank mRNA AK018178.1 - 7 - 8 628 147
AK020982.1 - 7 - 8 630 147
AK031683.1 - 6 - 7 534 147
AK032520.1 - 7 - 8 651 147
AK049888.1 - 7 - 8 627 147
AK079560.1 - 7 - 8 629 147

View Full Product Details