The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164827.1 | NP_001158299.1 | 2 - 3 | 586 | 88 |
NM_028054.3 | NP_082330.2 | 2 - 3 | 586 | 88 | |
GenBank mRNA | AK005386.1 | - | 2 - 3 | 350 | 88 |
BC018420.1 | - | 2 - 3 | 281 | 88 | |
BC099429.1 | - | 2 - 3 | 279 | 88 |