The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028085.2 | NP_082361.2 | 1 - 2 | 225 | 88 |
GenBank mRNA | AF524030.1 | - | 1 - 2 | 182 | 88 |
AK008229.1 | - | 1 - 2 | 207 | 88 | |
BC115784.1 | - | 1 - 2 | 186 | 88 | |
BC118013.1 | - | 1 - 2 | 186 | 88 | |
BY106969.1 | - | 1 - 2 | 227 | 88 |