Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdzd11
Entrez Gene ID
72621
Gene Name
PDZ domain containing 11
Gene Aliases
1810012H22Rik, 2310079D11Rik, 2700099C19Rik, AI854765, Pdzk11
Chromosome Location
Chr.X: 100622880 - 100625974 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028303.3 NP_082579.1 6 - 7 490 73
GenBank mRNA AK007454.1 - 6 - 7 444 73
AK012624.1 - 6 - 7 491 73
AK032327.1 - - 127 73
BC004608.1 - 6 - 7 448 73
BC022181.1 - 5 - 6 953 73

View Full Product Details