The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029537.1 | NP_083813.1 | 2 - 3 | 227 | 70 |
GenBank mRNA | AK148132.1 | - | 2 - 3 | 226 | 70 |
AK153960.1 | - | 2 - 3 | 217 | 70 | |
BC011208.1 | - | 2 - 3 | 227 | 70 |