The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007950.2 | NP_031976.1 | 1 - 2 | 183 | 66 |
GenBank mRNA | BB612312.1 | - | 1 - 2 | 178 | 66 |
BC027838.1 | - | 1 - 2 | 159 | 66 | |
CJ056327.1 | - | 1 - 2 | 184 | 66 | |
D30782.1 | - | 1 - 2 | 178 | 66 |