Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grid2
Entrez Gene ID
14804
Gene Name
glutamate receptor, ionotropic, delta 2
Gene Aliases
B230104L07Rik, GluD2, GluRdelta2, Lc, Lc<J>, MMS10-AC, Ms10ac, cpr, ho, nmf408, tpr
Chromosome Location
Chr.6: 63256832 - 64704322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008167.2 NP_032193.1 8 - 9 1246 86
GenBank mRNA AK036192.1 - 8 - 9 1277 86
BC139823.1 - 8 - 9 1250 86
D13266.1 - 8 - 9 1246 86

View Full Product Details