The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008412.3 | NP_032438.2 | 2 - 2 | 215 | 63 |
GenBank mRNA | AA789977.1 | - | 2 - 2 | 195 | 63 |
BC152326.1 | - | 2 - 2 | 195 | 63 | |
BY142984.1 | - | 2 - 2 | 217 | 63 |