The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190717.1 | NP_001177646.1 | 1 - 2 | 251 | 79 |
NM_013726.3 | NP_038754.1 | 1 - 2 | 251 | 79 | |
GenBank mRNA | AJ003132.1 | - | 1 - 2 | 141 | 79 |
AK014480.1 | - | 1 - 2 | 217 | 79 | |
AK132403.1 | - | 1 - 2 | 216 | 79 | |
BB611145.1 | - | 1 - 2 | 252 | 79 | |
BC050827.1 | - | 1 - 2 | 229 | 79 | |
BC069983.1 | - | 1 - 2 | 199 | 79 | |
BC138709.1 | - | 1 - 2 | 69 | 79 | |
BC138710.1 | - | 1 - 2 | 69 | 79 | |
BC145392.1 | - | 1 - 2 | 69 | 79 |