The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190448.1 | NP_001177377.1 | 2 - 3 | 345 | 132 |
NM_016672.4 | NP_057881.1 | 2 - 3 | 283 | 132 | |
GenBank mRNA | AF012881.1 | - | 2 - 3 | 233 | 132 |
AF012882.1 | - | 1 - 2 | 213 | 132 | |
AF071068.1 | - | 1 - 2 | 216 | 132 | |
AK011834.1 | - | 2 - 3 | 277 | 132 | |
AK149141.1 | - | 2 - 3 | 277 | 132 | |
AK160769.1 | - | 2 - 3 | 277 | 132 | |
BC060989.1 | - | 2 - 3 | 301 | 132 | |
BC089594.1 | - | 2 - 3 | 287 | 132 |