Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cldn7
Entrez Gene ID
53624
Gene Name
claudin 7
Gene Aliases
-
Chromosome Location
Chr.11: 69964779 - 69967886 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193619.1 NP_001180548.1 4 - 5 651 87
NM_016887.6 NP_058583.1 3 - 4 973 87
GenBank mRNA AF087825.1 - 3 - 4 467 87
AK002924.1 - 3 - 4 894 87
AK010558.1 - 3 - 4 893 87
AK087296.1 - 3 - 4 891 87
AK145504.1 - 3 - 4 974 87
AK167250.1 - 3 - 4 894 87
BC008104.1 - 3 - 4 538 87
BC050007.1 - 3 - 4 886 87
BG083098.2 - 1 - 2 94 87

View Full Product Details