The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018818.2 | NP_061288.2 | 11 - 12 | 1469 | 99 |
GenBank mRNA | AF218084.1 | - | 11 - 12 | 1478 | 99 |
AK035956.1 | - | 11 - 12 | 1464 | 99 | |
AK141830.1 | - | 10 - 11 | 1347 | 99 | |
BC047461.1 | - | 11 - 12 | 1468 | 99 |